missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ EIPA

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
TRPP3 channel inhibitor (IC50 = 10.5 μM). Inhibits the Na+/H+ exchanger (NHE). Derivative of amiloride (Cat. No. 0890).
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 299.76 |
| Purity | >98% |
| Molecular Weight (g/mol) | 299.76 |
| Chemical Name or Material | EIPA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction