missing translation for 'onlineSavingsMsg'
Learn More

EIPA hydrochloride, MedChemExpress

Product Code. 16496327
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16496327 5 mg 5mg
16423608 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496327 Supplier Medchem Express Supplier No. HY101840A
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress EIPA (L593754) hydrochloride is an orally active TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA hydrochloride also enhances autophagy by inhibiting Na+/H+-exchanger 3 (NHE3). EIPA hydrochloride inhibits macropinocytosis as well. EIPA hydrochloride can be used in the research of inflammation and cancers, such as gastric cancer, colon carcinoma, pancreatic carcinoma.

EIPA (L593754) hydrochloride is an orally active TRPP3 channel inhibitor with an IC50 of 10.5 μM. EIPA hydrochloride also enhances autophagy by inhibiting Na+/H+-exchanger 3 (NHE3). EIPA hydrochloride inhibits macropinocytosis as well. EIPA hydrochloride can be used in the research of inflammation and cancers, such as gastric cancer, colon carcinoma, pancreatic carcinoma.

Specifications

Chemical Name or Material EIPA hydrochloride
CAS 1345839-28-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C11H19Cl2N7O
Quantity 5 mg
Synonym L593754 hydrochloride MH 12-43 hydrochloride
Solubility Information DMSO : 130 mg/mL (386.65 mM; Need ultrasonic)
SMILES O=C(C1=NC(Cl)=C(N(CC)C(C)C)N=C1N)NC(N)=N.Cl
Molecular Weight (g/mol) 336.22
Formula Weight 336.22
Percent Purity 99.92%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.