missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIG121 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£164.00 - £375.00
Specifications
| Antigen | EIG121 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18340764
|
Novus Biologicals
NBP3-03728-20UL |
20 μL |
£164.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18344525
|
Novus Biologicals
NBP3-03728-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EIG121 Polyclonal specifically detects EIG121 in Mouse samples. It is validated for Western Blot.Specifications
| EIG121 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 57535 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| estrogen-induced gene 121 protein, hypothetical protein LOC57535, KIAA1324, MGC150624 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 42-300 of human EIG121 (NP_001253977.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title