missing translation for 'onlineSavingsMsg'
Learn More

EIG121 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18412397 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412397 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412397 Supplier Novus Biologicals Supplier No. NBP281949JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EIG121 Polyclonal antibody specifically detects EIG121 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIG121
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias estrogen-induced gene 121 protein, hypothetical protein LOC57535, KIAA1324, MGC150624
Host Species Rabbit
Immunogen EIG121 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human EIG121. The immunogen is located within the last 50 amino acids of EIG121. Amino Acid Squence: IKSFTSKRTPDGFDSVPL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 57535
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.