missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF5B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106663
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody detects endogenous levels of total EIF5B.
Accurate initiation of translation in eukaryotes is complex and requires many factors, some of which are composed of multiple subunits. The process is simpler in prokaryotes which have only three initiation factors (IF1, IF2, IF3). Two of these factors are conserved in eukaryotes: the homolog of IF1 is eIF1A and the homolog of IF2 is eIF5B. This gene encodes eIF5B. Factors eIF1A and eIF5B interact on the ribosome along with other initiation factors and GTP to position the initiation methionine tRNA on the start codon of the mRNA so that translation initiates accurately.
Spezifikation
| eIF5B | |
| Polyclonal | |
| Unconjugated | |
| EIF5B | |
| A030003E17Rik; Annexin V-binding protein ABP-7; BC018347; DKFZp434I036; eIF 5B; Eif5b; eIF-5B; Eukaryotic translation initiation factor 5B; FLJ10524; If2; KIAA0741; translation initiation factor IF2; Translation initiation factor IF-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 226982, 308306, 9669 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B2GUV7, O60841, Q05D44 | |
| EIF5B | |
| A synthesized peptide derived from human EIF5B(Accession O60841), corresponding to amino acid residues G1053-N1103. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur