missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539166
This item is not returnable.
View return policy
Description
Accurate initiation of translation in eukaryotes is complex and requires many factors, some of which are composed of multiple subunits. The process is simpler in prokaryotes which have only three initiation factors (IF1, IF2, IF3). Two of these factors are conserved in eukaryotes: the homolog of IF1 is eIF1A and the homolog of IF2 is eIF5B. This gene encodes eIF5B. Factors eIF1A and eIF5B interact on the ribosome along with other initiation factors and GTP to position the initiation methionine tRNA on the start codon of the mRNA so that translation initiates accurately.
Specifications
| EIF5B | |
| Polyclonal | |
| Unconjugated | |
| EIF5B | |
| Eukaryotic translation initiation factor 5B, Translation initiation factor IF2, IF2, FLJ10524, KIAA0741, DKFZp434I036, Translation initiation factor IF 2, eIF 5B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 226982, 308306, 9669 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60841, Q05D44, B2GUV7 | |
| EIF5B | |
| A synthetic peptide derived from the internal region of human EIF5B | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction