missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
eIF5A2 Monoclonal specifically detects eIF5A2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | eIF5A2 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 2 |
| Conjugate | DyLight 594 |
| Dilution | ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | eIF-5A-2, EIF-5A2, eIF5AII, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 2, eukaryotic translation initiation factor 5A2, eukaryotic translation initiation factor 5A-2 |
| Host Species | Rabbit |
| Immunogen | This antibody was obtained from a rabbit immunized with purified, recombinant Human eIF5A2 (Q9GZV4; Met1-Lys153). |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?