missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5A Rabbit anti-Human, Mouse, Rat, Clone: 3R10I7, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | eIF5A |
|---|---|
| Clone | 3R10I7 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18374722
|
Bio-Techne
NBP3-16409-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18310885
|
Novus Biologicals
NBP3-16409-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF5A Monoclonal antibody specifically detects eIF5A in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| eIF5A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1984 | |
| IgG | |
| Affinity purified |
| 3R10I7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| eIF-4D, eIF-5A, eIF-5A1, eIF-5A-1, EIF5A1, eIF5AI, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 1, eukaryotic translation initiation factor 5A, eukaryotic translation initiation factor 5A-1, MGC104255, MGC99547, Rev-binding factor | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human eIF5A (P63241). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title