missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5A Mouse anti-Human, Clone: 4D1, Novus Biologicals™
Mouse Monoclonal Antibody
£241.00 - £378.00
Specifications
| Antigen | eIF5A |
|---|---|
| Clone | 4D1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Flow Cytometry 1:10-1:1015, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18258072
|
Novus Biologicals
NBP2-59403-50UL |
50 μL |
£241.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18215671
|
Novus Biologicals
NBP2-59403-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF5A Monoclonal specifically detects eIF5A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| eIF5A | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| eIF-4D, eIF-5A, eIF-5A1, eIF-5A-1, EIF5A1, eIF5AI, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 1, eukaryotic translation initiation factor 5A, eukaryotic translation initiation factor 5A-1, MGC104255, MGC99547, Rev-binding factor | |
| EIF5A | |
| IgG1 κ | |
| Protein G purified |
| 4D1 | |
| Western Blot 1:500, Flow Cytometry 1:10-1:1015, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 1984 | |
| Recombinant human EIF5A (1-154aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title