Learn More
Invitrogen™ eIF5A Monoclonal Antibody (4E10G8)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA1179
Product Code. 15618018
  • £421.00 / 100µg
Description
MA1-179 detects EIF5A from human samples. MA1-179 has been successfully used in western blot, immunofluorescence, and flow cytometry applications.
mRNA-binding protein involved in translation elongation. Has an important function at the level of mRNA turnover, probably acting downstream of decapping. Involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintenance of cell wall integrity. With syntenin SDCBP, functions as a regulator of TP53/p53 and TP53/p53-dependent apoptosis. Regulates also TNF-alpha-mediated apoptosis. Mediates effects of polyamines on neuronal process extension and survival. May play an important role in brain development and function, and in skeletal muscle stem cell differentiation. Also described as a cellular cofactor of human T-cell leukemia virus type I (HTLV-1) Rex protein and of human immunodeficiency virus type 1 (HIV-1) Rev protein, essential for mRNA export of retroviral transcripts.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
eIF5A
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide; pH 7.4
P63241
EIF5A
Purified recombinant, full length human EIF5A (aa1-154) expressed in E. Coli.
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot, Immunocytochemistry
4E10G8
Unconjugated
EIF5A
AA410058; D19Wsu54e; Eif4d; eIF-4D; Eif5a; eIF-5A; EIF5A1; eIF-5A1; eIF-5A-1; eIF5AI; eukaryotic initiation factor 5A; Eukaryotic initiation factor 5A isoform 1; eukaryotic translation initiation factor 5A; eukaryotic translation initiation factor 5A-1; MGC104255; MGC99547; OTTHUMP00000128384; Rev-binding factor; uORF A
Mouse
Protein A
RUO
1984
-20°C, Avoid Freeze/Thaw Cycles