missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF5A Antibody (4D1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59403PE
This item is not returnable.
View return policy
Description
eIF5A Monoclonal antibody specifically detects eIF5A in Human samples. It is validated for Flow Cytometry
Specifications
| eIF5A | |
| Monoclonal | |
| PE | |
| eIF-4D, eIF-5A, eIF-5A1, eIF-5A-1, EIF5A1, eIF5AI, eukaryotic initiation factor 5A, Eukaryotic initiation factor 5A isoform 1, eukaryotic translation initiation factor 5A, eukaryotic translation initiation factor 5A-1, MGC104255, MGC99547, Rev-binding factor | |
| Recombinant human eIF5A (1-154aa) purified from E. coli | |
| 0.1 mL | |
| Apoptosis, Cell Biology | |
| 1984 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| 4D1 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction