Learn More
Invitrogen™ eIF5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117058
Description
Positive Samples: MCF7, SW480, HeLa, U-251MG Immunogen sequence: MSVNVNRSVS DQFYRYKMPR LIAKVEGKGN GIKTVIVNMV DVAKALNRPP TYPTKYFGCE LGAQTQFDVK NDRYIVNGSH EANKLQDMLD GFIKKFVLCP ECENPETDLH VNPKKQTIGN SCKACGYRGM LDTHHKLCTF ILKNPPENSD SGTGKKEKEK KNRKGKDKEN GSVSSSETPP PPPPPNEINP PPHTMEEEED DDWGEDTTEE AQRRRMDEIS DHAKVLTLSD DLERT.
Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
Specifications
| eIF5 | |
| Polyclonal | |
| Unconjugated | |
| Eif5 | |
| 2810011H21Rik; D12Ertd549e; EIF5; EIF-5; EIF-5A; eukaryotic initiation factor 5 (eIF-5); eukaryotic translation initiation factor 5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 108348073, 1983, 217869, 56783 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.91 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P55010, P59325, Q07205 | |
| Eif5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-235 of human EIF5 (NP_8921162). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.