missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF5 Monoclonal Antibody (6D6G6)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531727

Product Code. 16395154

  • £429.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-31727 has been tested in indirect ELISA.

Probable E3 ubiquitin-protein ligase which accepts ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfers the ubiquitin to targeted substrates. May be required for development of the head mesenchyme and neural tube closure.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

eIF5
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P55010
Eif5
Purified recombinant fragment of human EIF5 (AA: 1-300) expressed in E. coli.
100 μL
Primary
Human, Monkey
Antibody
IgG2a
ELISA, Western Blot
6D6G6
Unconjugated
Eif5
2810011H21Rik; D12Ertd549e; EIF5; EIF-5; EIF-5A; eukaryotic initiation factor 5 (eIF-5); eukaryotic translation initiation factor 5
Mouse
Protein G
RUO
1983
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.