missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4H Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541898
This item is not returnable.
View return policy
Description
Peptide sequence: DSRDDFNSGF RDDFLGGRGG SRPGDRRTGP PMGSRFRDGP PLRGSNMDFR Sequence homology: Dog: 100%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 100%; Rat: 93%.
This gene encodes one of the translation initiation factors, which functions to stimulate the initiation of protein synthesis at the level of mRNA utilization. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene generates 2 transcript variants.
Specifications
| eIF4H | |
| Polyclonal | |
| Unconjugated | |
| EIF4H | |
| AU018978; D5Ertd355e; E430026L18Rik; Ef4h; EIF4H; eIF-4H; Eukaryotic translation initiation factor 4H; KIAA0038; mKIAA0038; WBSCR1; Williams-Beuren syndrome chromosomal region 1 protein; Williams-Beuren syndrome chromosomal region 1 protein homolog; Williams-Beuren syndrome chromosome region 1; Williams-Beuren syndrome chromosome region 1 homolog; WSCR1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 7458 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q15056 | |
| EIF4H | |
| Synthetic peptide directed towards the middle region of human WBSCR1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction