missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4H Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57248
This item is not returnable.
View return policy
Description
eIF4H Polyclonal specifically detects eIF4H in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| eIF4H | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| eukaryotic translation initiation factor 4H, KIAA0038eIF-4H, WBSCR1, Williams-Beuren syndrome chromosome region 1, WSCR1Williams-Beuren syndrome chromosomal region 1 protein | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Human: 100%; Mouse: 92%; Rat: 85%; Trichomonas vaginalis: 84%; Chicken: 84%; Western clawed frog: 81%; Xenopus: 81%; Alistipes sp. HGB5: 76%;. | |
| Human, Mouse, Rat, Pig, Canine, Equine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q15056 | |
| EIF4H | |
| Synthetic peptides corresponding to WBSCR1 The peptide sequence was selected from the middle region of WBSCR1. Peptide sequence DSRDDFNSGFRDDFLGGRGGSRPGDRRTGPPMGSRFRDGPPLRGSNMDFR. | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 7458 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction