missing translation for 'onlineSavingsMsg'
Learn More

eIF4GII Antibody, Novus Biologicals™

Product Code. 18418950 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418950 25 μL 25µL
18498170 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18418950 Supplier Novus Biologicals Supplier No. NBP18486725ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

eIF4GII Polyclonal antibody specifically detects eIF4GII in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4GII
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol
Gene Alias eIF-4G 3, eIF-4-gamma 3, eIF-4-gamma II, eIF4GIIeIF4G 3, eukaryotic translation initiation factor 4 gamma 3, eukaryotic translation initiation factor 4 gamma, 3
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: EFDSRRTLTSRGSMGREKNDKPLPSATARPNTFMRGGSSKDLLDNQSQEEQRREMLETVKQLTGGVDVERNSTEAERNKTRESAKPEISAMSA
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8672
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.