Learn More
EIF4G1 Mouse anti-Human, Unconjugated, Clone: 2B10G8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67199-1-IG-20UL
 View more versions of this product
Product Code. 16869705
  • £135.00 / 20µL
Description
The EIF4G1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human EIF4G1. This antibody recognizes human, mouse, rat antigen. The EIF4G1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
EIF4G1
Monoclonal
500 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunofluorescence, ELISA
2B10G8
Unconjugated
20 μL
1981
IgG1