missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4G1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | eIF4G1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232657
|
Novus Biologicals
NBP3-33508-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30231690
|
Novus Biologicals
NBP3-33508-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF4G1 Monoclonal antibody specifically detects eIF4G1 in Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)Specifications
| eIF4G1 | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway, Phospho Specific | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 1981 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| DKFZp686A1451, EIF4F, eIF-4G 1, EIF-4G1, EIF4-gamma, eIF-4-gamma 1, EIF4GI, EIF4Gp220, eucaryotic translation initiation factor 4G, eukaryotic translation initiation factor 4 gamma 1, eukaryotic translation initiation factor 4 gamma, 1, P220 | |
| A synthetic peptide corresponding to a sequence within amino acids 551-650 of human eIF4G1 (NP_937884.2).,, Sequence:, GSNPGPESEGSGVPPRPEEADETWDSKEDKIHNAENIQPGEQKYEYKSDQWKPLNLEEKKRYDREFLLGFQFIFASMQKPEGLPHISDVVLDKANKTPLR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title