missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4G Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA596502
This item is not returnable.
View return policy
Description
Immunogen sequence: PEELLNGAPS PPAVDLSPVS EPEEQAKEVT ASMAPPTIPS ATPATAPSAT SPAQEEEMEE EEEEEEGEAG EAGEAESEKG GEELLPPEST PIPANLSQNL E; Positive Samples: MCF7, U251, BT474, HeLa
The protein encoded by this gene is a component of the protein complex EIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure, and recruitment of mRNA to the ribosome. Alternative splicing results in five transcript variants encoding four distinct isoforms.
Specifications
| eIF4G | |
| Polyclonal | |
| Unconjugated | |
| EIF4G1 | |
| DKFZp686A1451, E030015G23Rik, EIF4F, EIF4G, eIF-4G 1, EIF4G1, eIF-4G1, EIF4-gamma, eIF-4-gamma 1, EIF4GI, eIF4GI protein, eucaryotic translation initiation factor 4G, Eukaryotic translation initiation factor 4 gamma 1, eukaryotic translation initiation factor 4 gamma, 1, eukaryotic translation initiation factor 4, gamma 1, OTTHUMP00000210577, OTTHUMP00000210586, OTTHUMP00000210587, OTTHUMP00000210588, OTTHUMP00000210595, OTTHUMP00000210602, OTTHUMP00000210603, P220, PARK18 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1981 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.82 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide, pH 7.3 | |
| Q04637 | |
| EIF4G1 | |
| A synthetic peptide corresponding to a sequence within amino acids 400-500 of human EIF4G1 (NP_8865533) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction