missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4G Monoclonal Antibody (G.689.9)

Rabbit Monoclonal Antibody
Brand: Invitrogen™ MA514971
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
The protein encoded by this gene is a component of the protein complex EIF4F, which is involved in the recognition of the mRNA cap, ATP-dependent unwinding of 5'-terminal secondary structure, and recruitment of mRNA to the ribosome. Alternative splicing results in five transcript variants encoding four distinct isoforms.
Specifications
| eIF4G | |
| Monoclonal | |
| 215 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5 | |
| Q04637, Q6NZJ6 | |
| EIF4G1 | |
| Synthetic peptide corresponding to residues surrounding Gly188 of human eIF4G. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, RNA Immunoprecipitation, Immunocytochemistry | |
| G.689.9 | |
| Unconjugated | |
| EIF4G1 | |
| DKFZp686A1451; E030015G23Rik; EIF4F; EIF4G; eIF-4G 1; EIF4G1; eIF-4G1; EIF4-gamma; eIF-4-gamma 1; EIF4GI; eIF4GI protein; eucaryotic translation initiation factor 4G; Eukaryotic translation initiation factor 4 gamma 1; eukaryotic translation initiation factor 4 gamma, 1; eukaryotic translation initiation factor 4, gamma 1; OTTHUMP00000210577; OTTHUMP00000210586; OTTHUMP00000210587; OTTHUMP00000210588; OTTHUMP00000210595; OTTHUMP00000210602; OTTHUMP00000210603; P220; PARK18 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1981, 208643, 287986 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction