missing translation for 'onlineSavingsMsg'
Learn More

eIF4ENIF1 Antibody, Novus Biologicals™

Product Code. 18324899 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18324899 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324899 Supplier Novus Biologicals Supplier No. H00056478B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

eIF4ENIF1 Polyclonal antibody specifically detects eIF4ENIF1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Dual RNAscope ISH-IHC
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4ENIF1
Applications Western Blot, Immunocytochemistry, Immunoprecipitation, Dual RNAScope ISH-IHC
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Dual RNAscope ISH-IHC
Formulation PBS (pH 7.4)
Gene Accession No. AAH33028.1
Gene Alias 4E-TFLJ26551,2610509L04Rik, Clast4, eIF4E-transporter, eukaryotic translation initiation factor 4E nuclear import factor 1eIF4E transporter, eukaryotic translation initiation factor 4E transporter, FLJ21601
Host Species Mouse
Immunogen EIF4ENIF1 (AAH33028.1, 1 a.a. - 985 a.a.) full-length human protein. MDRRSMGETESGDAFLDLKKPPASKCPHRYTKEELLDIKELPHSKQRPSCLSEKYDSDGVWDPEKWHASLYPASGRSSPVESLKKELDTDRPSLVRRIVDPRERVKEDDLDVVLSPQRRSFGGGCHVTAAVSSRRSGSPLEKDSDGLRLLGGRRIGSGRIISARTFEKDHRLSDKDLRDLRDRDRERDFKDKRFRREFGDSKRVFGERRRNDSYTEEEPEWFSAGPTSQSETIELTGFDDKILEEDHKGRKRTRRRTASVKEGIVECNGGVAEEDEVEVILAQEPAADQEVPRDAVLPEQSPGDFDFNEFFNLDKVPCLASMIEDVLGEGSVSASRFSRWFSNPSRSGSRSSSLGSTPHEELERLAGLEQAILSPGQNSGNYFAPIPLEDHAENKVDILEMLQKAKVDLKPLLSSLSANKEKLKESSHSGVVLSVEEVEAGLKGLKVDQQVKNSTPFMAEHLEETLSAVTNNRQLKKDGDMTAFNKLVSTMKASGTLPSQPKVSRNLESHLMSPAEIPGQPVPKNILQELLGQPVQRPASSNLLSGLMGSLEPTTSLLGQRAPSPPLSQVFQTRAASADYLRPRIPSPIGFTPGPQQLLGDPFQGMRKPMSPITAQMSQLELQQAALEGLALPHDLAVQAANFYQPGFGKPQVDRTRDGFRNRQQRVTKSPAPVHRGNSSSPAPAASITSMLSPSFTPTSVIRKMYESKEKSKEEPASGKAALGDSKEDTQKASEENLLSSSSVPSADRDSSPTTNSKLSALQRSSCSTPLSQANRYTKEQDYRPKATGRKTPTLASPVPTTPFLRPVHQVPLVPHVPMVRPAHRLHPGLVQRMLAQGVHPQHLPSLLQTGVLPPGMDLSHLQGISGPILGQPFYPLPAASHPLLNPRPGTPLHLAMVQQQLQRSVLHPPGSGSHAAAVSVQTTPQNVPSRSGLPHMHSQLEHRPSQRSSSPVGLAKWFGSDVLQQPLPSMPAKVISVDELEYRQ
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline DNA Repair, DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 56478
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.