missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4EBP3 Antibody (OTI3B9), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71383R
This item is not returnable.
View return policy
Description
EIF4EBP3 Monoclonal specifically detects EIF4EBP3 in Human samples. It is validated for Western Blot.
Specifications
| EIF4EBP3 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| EIF4EBP3 | |
| Full length recombinant protein of human EIF4EBP3 (NP_003723) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| OTI3B9 | |
| Western Blot | |
| eIF4E-binding protein 3, eukaryotic translation initiation factor 4E binding protein 3,4E-BP3eukaryotic initiation factor 4E-binding protein 3, eukaryotic translation initiation factor 4E-binding protein 3,4EBP3 | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 8637.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction