missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4E3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17282-1-AP-20UL
This item is not returnable.
View return policy
Description
EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.[supplied by OMIM, Mar 2008].Specifications
| EIF4E3 | |
| Polyclonal | |
| Unconjugated | |
| EIF4E3 | |
| eIF 4E type 3, eIF 4E3, eIF4E 3, eIF4E type 3, EIF4E3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 297481, 317649, 66892 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N5X7, Q9DBB5 | |
| Eif4e3 | |
| EIF4E3 Fusion Protein Ag11056 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction