missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4E3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589499
This item is not returnable.
View return policy
Description
Immunogen sequence: MRGERRPLWE EESNAKGGVW KMKVPKDSTS TVWKELLLAT IGEQFTDCAA ADDEVIGVSV SVRDREDVVQ VWNVNASLVG EATVLEKIYE LLPHITFKAV FYKPHEEHHA FEGGRGKH; Positive Samples: Mouse lung, Rat lung.
EIF4E3 belongs to the EIF4E family of translational initiation factors that interact with the 5-prime cap structure of mRNA and recruit mRNA to the ribosome.
Specifications
| eIF4E3 | |
| Polyclonal | |
| Unconjugated | |
| Eif4e3 | |
| 1300018P11Rik; AI451927; eIF4E type 3; eIF-4E type 3; Eif4e3; eIF-4E3; eIF4E-3; eukaryotic translation initiation factor 4E class III; eukaryotic translation initiation factor 4E family member 3; eukaryotic translation initiation factor 4E member 3; eukaryotic translation initiation factor 4E type 3; eukaryotic translation initiation factor-3; wu:fc31f11; wu:fd15f11; zgc:92189; zgc:92189 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 297481, 66892 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.71 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9DBB5 | |
| Eif4e3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-118 of human EIF4E3 (NP_7754951). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction