missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4E3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12463
This item is not returnable.
View return policy
Description
EIF4E3 Polyclonal antibody specifically detects EIF4E3 in Drosophila samples. It is validated for Western Blot, ELISA
Specifications
| EIF4E3 | |
| Polyclonal | |
| Western Blot 1:500, ELISA 1:10000 | |
| eIF4E type 3, eIF-4E type 3, eIF-4E3, eIF4E-3, eukaryotic translation initiation factor 4E family member 3, eukaryotic translation initiation factor 4E member 3, eukaryotic translation initiation factor 4E type 3, MGC39820, MGC86971 | |
| Synthetic peptide taken within amino acid region 1-90 on eIF4E3 protein. | |
| 100 μg | |
| Primary | |
| Drosophila | |
| Purified |
| Western Blot, ELISA | |
| Unconjugated | |
| Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| 317649 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction