Learn More
Invitrogen™ eIF4E2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116997
Description
Positive Samples: BT-474, HL-60, Mouse kidney Immunogen sequence: MNNKFDALKD DDSGDHDQNE ENSTQKDGEK EKTERDKNQS SSKRKAVVPG PAEHPLQYNY TFWYSRRTPG RPTSSQSYEQ NIKQIGTFAS VEQFWRFYSH MVRPGDLTGH SDFHLFKEGI KPMWEDDANK NGGKWIIRLR KGLASRCWEN LILAMLGEQF MVGEEICGAV VSVRFQEDII SIWNKTASDQ ATTARIRDTL RRVLNLPPNT IMEYKTHTDS IKMPGRLGPQ RLLFQNLWKP.
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation (PubMed:9582349, PubMed:17368478, PubMed:25624349). Acts as a repressor of translation initiation (PubMed:22751931). In contrast to EIF4E, it is unable to bind eIF4G (EIF4G1, EIF4G2 or EIF4G3), suggesting that it acts by competing with EIF4E and block assembly of eIF4F at the cap. [UniProt].
Specifications
| eIF4E2 | |
| Polyclonal | |
| Unconjugated | |
| EIF4E2 | |
| 2700069E09Rik; 4EHP; 4E-LP; AI036339; AV129531; D0H0S6743E; EGK_04904; eIF4E type 2; eIF-4E type 2; Eif4e2; eIF4E-2; EIF4EL3; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; eukar; eukaryotic translation initiation factor 4E family member 2; eukaryotic translation initiation factor 4E homologous protein; eukaryotic translation initiation factor 4E like 3; eukaryotic translation initiation factor 4E member 2; eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; IF4e; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 2; mRNA cap-binding protein type 3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26987, 363275, 9470 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60573, Q8BMB3 | |
| EIF4E2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human EIF4E2 (NP_0048371). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.