missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4E2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110849
This item is not returnable.
View return policy
Description
Immunogen sequence: TERDKNQSSS KRKAVVPGPA EHPLQYNYTF WYSRRTPGRP TSSQSYEQNI KQIGTFASVE QFWRFYSHMV RPGDLTGHSD FHLFKEGIKP MWEDDANKNG GKWIIRL Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 95%.
Recognizes and binds the 7-methylguanosine-containing mRNA cap during an early step in the initiation (PubMed:9582349, PubMed:17368478, PubMed:25624349). Acts as a repressor of translation initiation (PubMed:22751931). In contrast to EIF4E, it is unable to bind eIF4G (EIF4G1, EIF4G2 or EIF4G3), suggesting that it acts by competing with EIF4E and block assembly of eIF4F at the cap. [UniProt].
Specifications
| eIF4E2 | |
| Polyclonal | |
| Unconjugated | |
| EIF4E2 | |
| 2700069E09Rik; 4EHP; 4E-LP; AI036339; AV129531; D0H0S6743E; EGK_04904; eIF4E type 2; eIF-4E type 2; Eif4e2; eIF4E-2; EIF4EL3; eIF4E-like cap-binding protein; eIF4E-like protein 4E-LP; eukar; eukaryotic translation initiation factor 4E family member 2; eukaryotic translation initiation factor 4E homologous protein; eukaryotic translation initiation factor 4E like 3; eukaryotic translation initiation factor 4E member 2; eukaryotic translation initiation factor 4E type 2; Eukaryotic translation initiation factor 4E-like 3; h4EHP; IF4e; mRNA cap-binding protein 4EHP; mRNA cap-binding protein type 2; mRNA cap-binding protein type 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9470 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O60573 | |
| EIF4E2 | |
| Recombinant Protein corresonding to Human eIF4E2. Recombinant protein control fragment (Product #RP-89635). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction