missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4E2 Antibody (OTI1F11), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71468PE
This item is not returnable.
View return policy
Description
EIF4E2 Monoclonal specifically detects EIF4E2 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| EIF4E2 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| EIF4E2 | |
| Full length recombinant protein of human EIF4E2 (NP_004837) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Flow Cytometry | |
| OTI1F11 | |
| Flow Cytometry | |
| 4EHP, 4E-LP, eIF4E type 2, eIF-4E type 2, EIF4EL3, eIF4E-like cap-binding protein, eIF4E-like protein 4E-LP, eukaryotic translation initiation factor 4E family member 2, Eukaryotic translation initiation factor 4E homologous protein, eukaryotic translation initiation factor 4E member 2, eukaryotic translation initiation factor 4E type 2, Eukaryotic translation initiation factor 4E-like 34E-LP, IF4e, mRNA cap-binding protein 4EHP, mRNA cap-binding protein type 3 | |
| Mouse | |
| Affinity Purified | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 9470.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction