missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4E Rabbit anti-Mouse, Rat, Clone: 7A7P2, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £386.00
Specifications
| Antigen | eIF4E |
|---|---|
| Clone | 7A7P2 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18369863
|
Novus Biologicals
NBP3-15629-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18327452
|
Novus Biologicals
NBP3-15629-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF4E Monoclonal antibody specifically detects eIF4E in Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| eIF4E | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 1977 | |
| IgG | |
| Affinity purified |
| 7A7P2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| CBP, eIF4E, eIF-4E, EIF4E1, EIF4EL1MGC111573, EIF4F, eIF-4F 25 kDa subunit, eukaryotic translation initiation factor 4E, eukaryotic translation initiation factor 4E-like 1, mRNA cap-binding protein | |
| A synthetic peptide corresponding to a sequence within amino acids 118-217 of human eIF4E (P06730). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title