missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4E, p Ser209 Antibody (SR1401), Novus Biologicals™
Rabbit Monoclonal Antibody
£265.00 - £341.00
Specifications
| Antigen | eIF4E |
|---|---|
| Clone | SR1401 |
| Dilution | Western Blot 1:1000-1:10000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18631864
|
Novus Biologicals
NBP3-21524-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18680555
|
Novus Biologicals
NBP3-21524-50ul |
50 μg |
£265.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
eIF4E Monoclonal antibody specifically detects eIF4E in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
| eIF4E | |
| Western Blot 1:1000-1:10000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CBP, eIF4E, eIF-4E, EIF4E1, EIF4EL1MGC111573, EIF4F, eIF-4F 25 kDa subunit, eukaryotic translation initiation factor 4E, eukaryotic translation initiation factor 4E-like 1, mRNA cap-binding protein | |
| A synthesized peptide derived from human phospho-eIF4E [Ser209] (Uniprot #: P06730 ) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR1401 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway, Phospho Specific | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 1977 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title