missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4E Antibody (5D11), Novus Biologicals™
Mouse Monoclonal Antibody
£184.00
Specifications
| Antigen | eIF4E |
|---|---|
| Clone | 5D11 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
Description
eIF4E Monoclonal specifically detects eIF4E in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| eIF4E | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 1977 | |
| Purified recombinant fragment of human eIF4E expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 5D11 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway, Phospho Specific | |
| CBP, eIF4E, eIF-4E, EIF4E1, EIF4EL1MGC111573, EIF4F, eIF-4F 25 kDa subunit, eukaryotic translation initiation factor 4E, eukaryotic translation initiation factor 4E-like 1, mRNA cap-binding protein | |
| EIF4E | |
| IgG1 | |
| Unpurified | |
| 25 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title