missing translation for 'onlineSavingsMsg'
Learn More

eIF4E Antibody (299910) [HRP], Novus Biologicals™

Product Code. 30735129 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735129 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735129 Supplier Novus Biologicals Supplier No. FAB3228H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

eIF4E Monoclonal antibody specifically detects eIF4E in Human,Mouse,Rat samples. It is validated for Western Blot,Immunocytochemistry,Gene Knock-Out
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4E
Applications Western Blot, Immunocytochemistry, Gene Knock-Out
Classification Monoclonal
Clone 299910
Conjugate HRP
Formulation PBS
Gene Alias CBP, eIF4E, eIF-4E, EIF4E1, EIF4EL1MGC111573, EIF4F, eIF-4F 25 kDa subunit, eukaryotic translation initiation factor 4E, eukaryotic translation initiation factor 4E-like 1, mRNA cap-binding protein
Host Species Mouse
Immunogen E. coli-derived recombinant human eIF4E, Met1-Val217, Accession # P06730
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, DNA replication Transcription Translation and Splicing, mTOR Pathway, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 1977
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.