missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF4A3 Recombinant Rabbit Monoclonal Antibody (K01_1Q69)

Product Code. 30828634
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828634 100 μL
30829543 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828634 Supplier Invitrogen™ Supplier No. MA563782

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

eIF4A3 Recombinant Monoclonal Antibody for ICC/IF, IHC (P), Western Blot

eIF4A3 is a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. eIF4A3 is a nuclear matrix protein. Its amino acid sequence is highly similar to the amino acid sequences of the translation initiation factors eIF4AI and eIF4AII, two other members of the DEAD box protein family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4A3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone K01_1Q69
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gene EIF4A3
Gene Accession No. P38919, Q3B8Q2
Gene Alias 2400003O03Rik; ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-3; Ddx48; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; Eif4a3; eIF4AIII; eIF-4A-III; eIF4A-III; eukaryotic initiation factor 4AIII; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-III, N-terminally processed; Eukaryotic initiation factor 4A-III-like protein; eukaryotic initiation factor 4A-like NUK-34; eukaryotic translation initiation factor 4A; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 3; eukaryotic translation initiation factor 4A3; hNMP 265; KIAA0111; mKIAA0111; MUK34; NMP 265; NMP265; Nuclear matrix protein 265; NUK34; RCPS; similar to DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; zgc:56139
Gene Symbols EIF4A3
Host Species Rabbit
Immunogen A synthetic peptide of human Eif4a3.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100757133, 688288, 9775
Target Species Hamster, Human, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.