missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF4A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554532
This item is not returnable.
View return policy
Description
Immunogen sequence: MATTATMATS GSARKRLLKE EDMTKVEFET SEEVDVTPTF DTMGLREDLL RGIYAYGFEK PSAIQQRAIK QIIK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 99%.
eIF4A3 is a member of the DEAD box protein family. DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure, such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. eIF4A3 is a nuclear matrix protein. Its amino acid sequence is highly similar to the amino acid sequences of the translation initiation factors eIF4AI and eIF4AII, two other members of the DEAD box protein family.
Specifikationer
| eIF4A3 | |
| Polyclonal | |
| Unconjugated | |
| EIF4A3 | |
| 2400003O03Rik; ATP-dependent RNA helicase DDX48; ATP-dependent RNA helicase eIF4A-3; Ddx48; DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; DEAD box protein 48; Eif4a3; eIF4AIII; eIF-4A-III; eIF4A-III; eukaryotic initiation factor 4AIII; Eukaryotic initiation factor 4A-III; Eukaryotic initiation factor 4A-III, N-terminally processed; Eukaryotic initiation factor 4A-III-like protein; eukaryotic initiation factor 4A-like NUK-34; eukaryotic translation initiation factor 4A; Eukaryotic translation initiation factor 4A isoform 3; eukaryotic translation initiation factor 4A, isoform 3; eukaryotic translation initiation factor 4A3; hNMP 265; KIAA0111; mKIAA0111; MUK34; NMP 265; NMP265; Nuclear matrix protein 265; NUK34; RCPS; similar to DEAD (Asp-Glu-Ala-Asp) box polypeptide 48; zgc:56139 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9775 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P38919 | |
| EIF4A3 | |
| Recombinant protein corresponding to Human eIF4A3. Recombinant protein control fragment (Product #RP-93301). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering