missing translation for 'onlineSavingsMsg'
Learn More

EIF4A3 Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18335103 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18335103 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18335103 Supplier Novus Biologicals Supplier No. NBP306400AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

EIF4A3 Polyclonal antibody specifically detects EIF4A3 in Human samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen EIF4A3
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias ATP-dependent RNA helicase DDX48, ATP-dependent RNA helicase eIF4A-3, DDX48, DEAD (Asp-Glu-Ala-Asp) box polypeptide 48, DEAD box protein 48, DKFZp686O16189, EC 3.6.1, EC 3.6.4.13, eIF-4A-III, eIF4A-III, EIF4AIII, eukaryotic initiation factor 4A-III, Eukaryotic initiation factor 4A-like NUK-34, eukaryotic translation initiation factor 4A, Eukaryotic translation initiation factor 4A isoform 3, eukaryotic translation initiation factor 4A3, hNMP 265, KIAA0111eukaryotic translation initiation factor 4A, isoform 3, MGC10862, NMP 265, NMP265, Nuclear matrix protein 265, NUK34
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human EIF4A3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 9775
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.