missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11280-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
eIF4A2 is a 407 amino acid ATP-dependent RNA helicase belonging to the DEAD box helicase family. It contains one helicase ATP-binding domain and a helicase C-terminal domain. eIF4A2, a subunit of the eIF4F complex, acts as a protein-synthesis initiation factor and thus plays an important role in the binding of mRNA to the 43S pre-initiation complex of ribosomes. Human eIF4A2 interacts with the NS5B protein of Hepatitis C virus (HCV) and facilitates the genomic RNA synthesis of NS5B protein by unwinding the secondary structure of the HCV genome. It is ubiquitously expressed in most of normal tissues, but in variable amounts.Especificaciones
| EIF4A2 | |
| Polyclonal | |
| Unconjugated | |
| EIF4A2 | |
| BM 010, DDX2B, eIF 4A II, EIF4A, eIF4A II, EIF4A2, EIF4F | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13682, 1974, 303831 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P10630, Q14240, Q5RKI1 | |
| EIF4A2 | |
| EIF4A2 Fusion Protein Ag1810 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido