missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF4A2 Mouse anti-Human, Mouse, Rat, CoraLite 594, Clone: 2D11E11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL594-67799-20UL
This item is not returnable.
View return policy
Description
EIF4A2 Monoclonal antibody specifically detects EIF4A2 in Human, Mouse, Rat samples. It is validated for FC (Intra), IFSpecifications
| EIF4A2 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Mouse | |
| 20 μL | |
| 1974 | |
| Store at -20°C | |
| Liquid |
| Flow Cytometry, Immunofluorescence | |
| 2D11E11 | |
| CoraLite 594 | |
| BC015831 | |
| Protein A purification | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction