missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
eIF4A2 Monoclonal specifically detects eIF4A2 in Human samples. It is validated for ELISA, Protein Array.
Specifications
Specifications
| Antigen | eIF4A2 |
| Applications | ELISA, Peptide Array |
| Classification | Monoclonal |
| Clone | PCRP-EIF4A2-2B5 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | ELISA, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?