missing translation for 'onlineSavingsMsg'
Learn More

eIF4A2 Antibody (PCRP-EIF4A2-2B5) - Azide and BSA Free, Novus Biologicals™

Product Code. 18328672 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328672 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328672 Supplier Novus Biologicals Supplier No. NBP308883

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

eIF4A2 Monoclonal specifically detects eIF4A2 in Human samples. It is validated for ELISA, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF4A2
Applications ELISA, Peptide Array
Classification Monoclonal
Clone PCRP-EIF4A2-2B5
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA, Protein Array
Formulation 10mM PBS
Gene Alias ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2
Host Species Mouse
Immunogen Recombinant full-length human eIF4A2 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1974
Target Species Human
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.