missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF4A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-57716
This item is not returnable.
View return policy
Description
eIF4A2 Polyclonal specifically detects eIF4A2 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| eIF4A2 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| ATP-dependent RNA helicase eIF4A-2, BM-010, DDX2Beukaryotic translation initiation factor 4A, isoform 2, EC 3.6.1, EC 3.6.4.13, EIF4A, eIF-4A-II, eIF4A-II, EIF4F, eukaryotic initiation factor 4A-II, eukaryotic translation initiation factor 4A, eukaryotic translation initiation factor 4A2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| EIF4A2 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SNWNEIVDNFDDMNLKESLLRGIYA | |
| 100 μL | |
| Cancer, mTOR Pathway | |
| 1974 | |
| Human | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering