missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3k Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550032
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total EIF3K protein.
The Wilms tumor suppressor gene WT1 appears to play a role in both transcriptional and posttranscriptional regulation of certain cellular genes. This gene encodes a WT1-associating protein, which is a ubiquitously expressed nuclear protein. Like WT1 protein, this protein is localized throughout the nucleoplasm as well as in speckles and partially colocalizes with splicing factors. Alternative splicing of this gene results in three transcript variants, two of which encode the same isoform.
Specifications
| eIF3k | |
| Polyclonal | |
| Unconjugated | |
| EIF3K | |
| 1200009C21Rik; ARG134; eIF-3 p25; eIF-3 p28; eIF3k; EIF3-p28; Eif3s12; eukaryotic translation initiation factor 3 subunit 12; Eukaryotic translation initiation factor 3 subunit K; eukaryotic translation initiation factor 3, subunit 12; eukaryotic translation initiation factor 3, subunit K; HSPC029; M9; MSTP001; muscle specific; muscle-specific gene M9 protein; PLAC24; PLAC-24; PRO1474; PTD001 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27335 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UBQ5 | |
| EIF3K | |
| Synthesized peptide derived from internal of human EIF3K (aa 4-31). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction