missing translation for 'onlineSavingsMsg'
Learn More
Learn More
eIF3K Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31285
This item is not returnable.
View return policy
Description
eIF3K Polyclonal specifically detects eIF3K in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| eIF3K | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| ARG134, eIF3k, EIF3-p28, EIF3S12eIF-3 p28, Eukaryotic translation initiation factor 3 subunit 12, eukaryotic translation initiation factor 3 subunit K, eukaryotic translation initiation factor 3, subunit 12, eukaryotic translation initiation factor 3, subunit K, HSPC029, M9, MSTP001, muscle specific, Muscle-specific gene M9 protein, PLAC24, PLAC-24, PLAC-24eIF-3 p25, PRO1474, PTD001 | |
| Rabbit | |
| 25 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 27335 | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 4.65mg/mL | |
| Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
| Q9UBQ5 | |
| EIF3K | |
| Recombinant protein encompassing a sequence within the center region of human EIF3K. The exact sequence is proprietary. | |
| Protein A purified | |
| RUO | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Mouse(100%), Cow(100%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction