Learn More
eIF3K Antibody (01), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-06567UV
Product Code. 18559921
  • £329.00 / 0.10mL
Description
eIF3K Monoclonal antibody specifically detects eIF3K in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
eIF3K
Monoclonal
DyLight 350
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified
Immunofluorescence
01
Immunocytochemistry/Immunofluorescence
ARG134, eIF3k, EIF3-p28, EIF3S12eIF-3 p28, Eukaryotic translation initiation factor 3 subunit 12, eukaryotic translation initiation factor 3 subunit K, eukaryotic translation initiation factor 3, subunit 12, eukaryotic translation initiation factor 3, subunit K, HSPC029, M9, MSTP001, muscle specific, Muscle-specific gene M9 protein, PLAC24, PLAC-24, PLAC-24eIF-3 p25, PRO1474, PTD001
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human eIF3K (Q9UBQ5; Ala2-Gln218).
0.1 mL
Core ESC Like Genes, Stem Cell Markers
27335
Store at 4°C in the dark.