missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF3j Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA597138

Product Code. 16549971

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MAAAAAAAGD SDSWDADAFS VEDPVRKVGG GGTAGGDRWE GEDEDEDVKD NWDDDDDEKK EEAEVKPEVK ISEKKKIAEK IKEKERQQKK RQEEIKKRLE EPEEPKVLTP EEQLADKLRL KKLQEESDLE LAKETFGVNN AVYGIDAMNP SSRDDFTEFG KLLKDKITQY EKSLYYASFL EVLVRDVCIS LEIDDLKKIT NSLTVLCSEK QKQEKQSKAK KKKKGVVPGG GLKATMKDDL ADYGGYDGGY VQDYEDFM; Positive Samples: SKOV3, HeLa, Jurkat, A-549, U-87MG, LO2, Mouse testis, Mouse liver; Cellular Location: Cytoplasm.

Eukaryotic initiation factor-3 (EIF3) has a molecular mass of about 600 kD and contains 13 nonidentical protein subunits, including EIF3J. EIF3 plays a central role in binding of initiator methionyl-tRNA and mRNA to the 40S ribosomal subunit to form the 40S initiation complex (Fraser et al., 2004; Fraser et al., 2007).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

eIF3j
Polyclonal
Unconjugated
EIF3J
1810016I04Rik; 2700079K05Rik; AA409117; AA409446; C78575; eIF3 p35; eIF3 p35 {ECO:0000255; eIF-3-alpha; eIF3-alpha; eIF-3-alpha {ECO:0000255; eIF-3-alpha-A; eIF-3-alpha-B; Eif3j; eIF3j {ECO:0000255; Eif3j1; Eif3j-1; Eif3j2; Eif3j-2; eIF3j-A; eIF3j-B; eIF3-p35; EIF3S1; Eif3s1-1; Eif3s1-2; ENSMUSG00000043424; Eukaryotic translation initiation factor 3 subunit 1; eukaryotic translation initiation factor 3 subunit 1 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 1-A; Eukaryotic translation initiation factor 3 subunit 1-B; eukaryotic translation initiation factor 3 subunit J; eukaryotic translation initiation factor 3 subunit J {ECO:0000255; Eukaryotic translation initiation factor 3 subunit J-A; Eukaryotic translation initiation factor 3 subunit J-B; eukaryotic translation initiation factor 3, subunit 1 (alpha, 35kD); eukaryotic translation initiation factor 3, subunit 1 alpha; eukaryotic translation initiation factor 3, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 3, subunit J; eukaryotic translation initiation factor 3, subunit J1; eukaryotic translation initiation factor 3, subunit J2; Gm9781; HAMAP-Rule:MF_03009}; PRO0391
Rabbit
Affinity Chromatography
RUO
100042807, 691947, 78655, 8669
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.34 mg/mL
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
A0JPM9, O75822, Q3UGC7, Q66JS6
EIF3J, Eif3j1, Eif3j2
Recombinant protein (or fragment).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.