missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3i Monoclonal Antibody (3B10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA520471
This item is not returnable.
View return policy
Description
Peptide Sequence: FRTERPVNSA ALSPNYDHVV LGGGQEAMDV TTTSTRIGKF EARFFHLAFE EEFGRVKGHF GPINSVAFHP DGKSYSSGGE DGYVRIHYFD PQYFEFEFEA.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
Specifications
| eIF3i | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q13347 | |
| Eif3i | |
| EIF3I (NP_003748.1, 226 a.a. ~ 325 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 3B10 | |
| Unconjugated | |
| Eif3i | |
| 36kDa; D4Ertd632e; eIF3 p36; eIF-3-beta; eIF3-beta; Eif3i; eIF3-p36; Eif3s2; Eukaryotic translation initiation factor 3 subunit 2; eukaryotic translation initiation factor 3 subunit I; eukaryotic translation initiation factor 3, subunit 1; eukaryotic translation initiation factor 3, subunit 2 (beta); eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 2 beta, 36kDa; eukaryotic translation initiation factor 3, subunit I; predicted protein of HQ2242; PRO2242; subuni; TGF-beta receptor binding protein; TGF-beta receptor-interacting protein 1; TGFbeta receptor-interacting protein 1; TRIP1; TRIP-1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8668 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction