missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3h Monoclonal Antibody (GT35512)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527798
This item is not returnable.
View return policy
Description
Positive Control: A431 Predicted Reactivity: Rat (99%), Xenopus laevis (86%), Chicken (95%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
eIF3H is a component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S pre-initiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post-termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. The eIF-3 complex specifically targets and initiates translation of a subset of mRNAs involved in cell proliferation, including cell cycling, differentiation and apoptosis, and uses different modes of RNA stem-loop binding to exert either translational activation or repression.
Specifications
| eIF3h | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O15372, Q91WK2 | |
| Eif3h | |
| Recombinant protein encompassing a sequence within the center region of human eIF3H/EIF3S3. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| GT35512 | |
| Unconjugated | |
| Eif3h | |
| 1110008A16Rik; 40 kDa; 40kD; 9430017H16Rik; eIF-3 gamma; eIF3 p40 subunit; eIF3 p40 subunit {ECO:0000255; eIF-3-gamma; eIF3-gamma; EIF3H; eIF3h {ECO:0000255; eIF3p40; eIF3-p40; EIF3S3; eukaryotic translation initiation factor 3 subunit 3; eukaryotic translation initiation factor 3 subunit 3 {ECO:0000255; eukaryotic translation initiation factor 3 subunit 3 gamma; eukaryotic translation initiation factor 3 subunit H; eukaryotic translation initiation factor 3 subunit H {ECO:0000255; eukaryotic translation initiation factor 3 subunit H-like protein; eukaryotic translation initiation factor 3, subunit 2 (beta, 36kD); eukaryotic translation initiation factor 3, subunit 3 (gamma); eukaryotic translation initiation factor 3, subunit 3 (gamma, 40kD); eukaryotic translation initiation factor 3, subunit 3 gamma, 40kDa; eukaryotic translation initiation factor 3, subunit H; eukaryotic translation initiation factor subunit H-like protein; HAMAP-Rule:MF_03007}; MGC102958 | |
| Mouse | |
| Protein A | |
| RUO | |
| 68135, 8667 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction