missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3G Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | EIF3G |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18340027
|
Novus Biologicals
NBP3-04672-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18358636
|
Novus Biologicals
NBP3-04672-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EIF3G Polyclonal specifically detects EIF3G in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| EIF3G | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 8666 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| eIF3 p42, eIF3 p44, eIF-3-delta, eIF3-delta, eIF3g, EIF3-P42, eIF3-p44, EIF3S4eIF-3 RNA-binding subunit, Eukaryotic translation initiation factor 3 RNA-binding subunit, Eukaryotic translation initiation factor 3 subunit 4, eukaryotic translation initiation factor 3 subunit G, eukaryotic translation initiation factor 3 subunit p42, eukaryotic translation initiation factor 3, subunit 4 (delta, 44kD), eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa, eukaryotic translation initiation factor 3, subunit G | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-320 of human EIF3G (NP_003746.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title