missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eIF3g Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA523287

Product Code. 13227918

  • £442.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The amino acid sequence used as immunogen is 84% homologous in mouse.

Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of posttermination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. This subunit can bind 18S rRNA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

eIF3g
Polyclonal
Unconjugated
EIF3G
44kDa; D0Jmb4; eiF3 delta; eIF3 p42; eIF3 p42 {ECO:0000255; eIF3 p44; eIF3 p44 {ECO:0000255; eIF-3 RNA-binding subunit; eIF-3 RNA-binding subunit {ECO:0000255; eIF-3-delta; eIF3-delta; eIF-3-delta {ECO:0000255; EIF3G; eIF3g {ECO:0000255; Eif3p42; EIF3-P42; eIF3-p44; EIF3S4; eukaryotic translation initiation factor 3 RNA-binding subunit; eukaryotic translation initiation factor 3 RNA-binding subunit {ECO:0000255; Eukaryotic translation initiation factor 3 subunit 4; eukaryotic translation initiation factor 3 subunit 4 {ECO:0000255; eukaryotic translation initiation factor 3 subunit G; eukaryotic translation initiation factor 3 subunit G {ECO:0000255; eukaryotic translation initiation factor 3, subunit 4 (delta); eukaryotic translation initiation factor 3, subunit 4 (delta, 44 kDa); eukaryotic translation initiation factor 3, subunit 4 (delta, 44kD); eukaryotic translation initiation factor 3 subunit p42; eukaryotic translation initiation factor 3, subunit 4 delta; eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa; eukaryotic translation initiation factor 3, subunit G; HAMAP-Rule:MF_03006}; p44; similar to eukaryotic translation initiation factor 3, subunit 4 delta, 44kDa; translation initiation factor eIF3-p44; TU-189B2; Unknown (protein for MGC:128341); zgc:56553
Rabbit
Protein A
RUO
8666
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O75821
EIF3G
A portion of amino acids 50-100 of human eIF3G.
100 μg
Primary
Human, Monkey
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.