Learn More
Invitrogen™ eIF3f Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596549
Description
Immunogen sequence: GGRVVRLHPV ILASIVDSYE RRNEGAARVI GTLLGTVDKH SVEVTNCFSV PHNESEDEVA VDMEFAKNMY ELHKKVSPNE LILGWYATGH DITEHSVLIH EYYSREAPNP IHLTVDTSLQ NGRMSIKAYV STLMGVPGRT MGVMFTPLTV KYAYYDTERI GVDLIMKTCF SPNRVIGLSS DLQQVGGASA RIQDALSTVL QYAEDVLSGK VSADNTVGRF LMSLVNQVPK IVPDDFETML NSNINDLLMV TYLANLTQSQ IALNEKLVNL; Positive Samples: HL-60, SKOV3, BT-474, Jurkat, HeLa, Mouse spleen, Mouse lung, Mouse heart, Rat brain; Cellular Location: Cytoplasm.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation.
Specifications
| eIF3f | |
| Polyclonal | |
| Unconjugated | |
| EIF3F | |
| 0610037M02Rik; AA409853; deubiquitinating enzyme eIF3f; eIF3 p47; eIF-3-epsilon; eIF3-epsilon; EIF3F; eIF3-p47; Eif3s5; eukaryotic translation initiation factor 3 subunit 5; eukaryotic translation initiation factor 3 subunit 5 epsilon; eukaryotic translation initiation factor 3 subunit F; eukaryotic translation initiation factor 3, subunit 5 (epsilon); eukaryotic translation initiation factor 3, subunit 5 (epsilon, 47kD); eukaryotic translation initiation factor 3, subunit 5 epsilon, 47kDa; eukaryotic translation initiation factor 3, subunit F | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 293427, 66085, 8665 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 3.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O00303, Q9DCH4 | |
| EIF3F | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 88-357 of human EIF3F (NP_0037451). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.