missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eIF3e Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582859
This item is not returnable.
View return policy
Description
Immunogen sequence: ARLFIFETFC RIHQCISINM LADKLNMTPE EAERWIVNLI RNARLDAKID SKLGHVVMGN NAVSPYQQVI EKTKSLSFRS QMLAMNIEKK LNQNSRSEAP NWATQDSGFY Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of post termination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. Required for nonsense-mediated mRNA decay (NMD); may act in conjunction with UPF2 to divert mRNAs from translation to the NMD pathway. May interact with MCM7 and EPAS1 and regulate the proteasome-mediated degradation of these proteins.
Specifications
| eIF3e | |
| Polyclonal | |
| Unconjugated | |
| EIF3E | |
| 48kDa; eIF-3 p48; eIF3e; eIF3-p46; EIF3-P48; EIF3S6; eukaryotic translation initiation factor 3 subunit 6; Eukaryotic translation initiation factor 3 subunit E; eukaryotic translation initiation factor 3 subunit E isoform 2 transcript; eukaryotic translation initiation factor 3 subunit E-like protein; eukaryotic translation initiation factor 3, subunit 6 (48kD); eukaryotic translation initiation factor 3, subunit 6 48kDa; eukaryotic translation initiation factor 3, subunit E; eukaryotic translation initiation factor subunit E-like protein; Int6; mammary tumor integration site 6; mammary tumor-associated protein INT6; mammary tumor-associated protein INT-6; MMTV integration site 6; murine mammary tumor integration site 6 (oncogene homolog); NEWGENE_1311521; Viral integration site protein INT-6; viral integration site protein INT-6 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 103692906, 16341, 3646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P60228, P60229, Q641X8 | |
| EIF3E | |
| Recombinant protein corresponding to Human EIF3E. Recombinant protein control fragment (Product #RP-92457). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction